Assay Details
| Gene Symbol: PTEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123302.4 | NP_198756.2 | 5 - 6 | 895 | 72 | |
| GenBank mRNA | AJ490172.1 | - | 5 - 6 | 789 | 72 | |
| AK118115.1 | - | 5 - 6 | 877 | 72 | ||
| BT005560.1 | - | 4 - 5 | 759 | 72 | ||
More Information
Set Membership: |
|

PTEN1
Genomic Map