Assay Details
| Gene Symbol: PTEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123302.4 | NP_198756.2 | 7 - 8 | 1228 | 108 | |
| GenBank mRNA | AJ490172.1 | - | 7 - 8 | 1122 | 108 | |
| AK118115.1 | - | 7 - 8 | 1210 | 108 | ||
| BT005560.1 | - | 6 - 7 | 1092 | 108 | ||
More Information
Set Membership: |
|

PTEN1
Genomic Map