Assay Details
| Gene Symbol: CLPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123356.3 | NP_198809.2 | 2 - 3 | 349 | 124 | |
| GenBank mRNA | AY954872.1 | - | 2 - 3 | 250 | 124 | |
More Information
Set Membership: |
|

CLPS5
Genomic Map