Assay Details
| Gene Symbol: AGL76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123375.1 | NP_198828.1 | 1 | 965 | 99 | |
| GenBank mRNA | AY141222.1 | - | 1 | 965 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL76| Gene Symbol: AGL76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123375.1 | NP_198828.1 | 1 | 965 | 99 | |
| GenBank mRNA | AY141222.1 | - | 1 | 965 | 99 | |
Set Membership: |
|