Assay Details
| Gene Symbol: EMB3012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123412.4 | NP_198864.2 | 8 - 9 | 1955 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB3012| Gene Symbol: EMB3012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123412.4 | NP_198864.2 | 8 - 9 | 1955 | 105 | |
Set Membership: |
|