Assay Details
| Gene Symbol: G6PD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344352.1 | NP_001330361.1 | 15 - 16 | 1599 | 72 | |
| NM_123441.3 | NP_198892.1 | 15 - 16 | 1664 | 72 | ||
| GenBank mRNA | AK227918.1 | - | 15 - 16 | 1579 | 72 | |
| BT004633.1 | - | 14 - 15 | 1386 | 72 | ||
More Information
Set Membership: |
|

G6PD6
Genomic Map