Assay Details
| Gene Symbol: POLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123553.6 | NP_199003.5 | 4 - 5 | 451 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLA3| Gene Symbol: POLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123553.6 | NP_199003.5 | 4 - 5 | 451 | 102 | |
Set Membership: |
|