Assay Details
| Gene Symbol: AT5G42660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123630.5 | NP_568609.2 | 4 - 5 | 1204 | 68 | |
| GenBank mRNA | AY057714.1 | - | 4 - 5 | 973 | 68 | |
| AY124876.1 | - | 4 - 5 | 886 | 68 | ||
| KY906088.1 | - | 4 - 5 | 886 | 68 | ||
More Information
Set Membership: |
|

AT5G42660
Genomic Map