Assay Details
| Gene Symbol: UBC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123665.5 | NP_568619.1 | 3 - 4 | 319 | 101 | |
| GenBank mRNA | AY058137.1 | - | 3 - 4 | 271 | 101 | |
| AY101537.1 | - | 3 - 4 | 235 | 101 | ||
| DQ027032.1 | - | 3 - 4 | 235 | 101 | ||
More Information
Set Membership: |
|

UBC18
Genomic Map