Assay Details
| Gene Symbol: PUM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123677.2 | NP_199125.2 | 3 - 4 | 1202 | 68 | |
| GenBank mRNA | DQ056705.1 | - | 3 - 4 | 1202 | 68 | |
More Information
Set Membership: |
|

PUM14
Genomic Map