Assay Details
| Gene Symbol: AGL80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124244.3 | NP_199678.1 | 1 | 819 | 139 | |
| GenBank mRNA | DQ056710.1 | - | 1 | 768 | 139 | |
| DQ406752.1 | - | 1 | 819 | 139 | ||
More Information
Set Membership: |
|

AGL80
Genomic Map