Assay Details
| Gene Symbol: LPA66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124268.2 | NP_199702.1 | 1 | 1985 | 113 | |
| GenBank mRNA | AY056318.1 | - | 1 | 1771 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPA66| Gene Symbol: LPA66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124268.2 | NP_199702.1 | 1 | 1985 | 113 | |
| GenBank mRNA | AY056318.1 | - | 1 | 1771 | 113 | |
Set Membership: |
|