Assay Details
| Gene Symbol: MAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124463.4 | NP_199898.1 | 10 - 11 | 981 | 111 | |
| GenBank mRNA | AY070728.1 | - | 10 - 11 | 816 | 111 | |
| BT000839.1 | - | 9 - 10 | 688 | 111 | ||
More Information
Set Membership: |
|

MAB1
Genomic Map