Assay Details
| Gene Symbol: MAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124463.4 | NP_199898.1 | 12 - 13 | 1096 | 59 | |
| GenBank mRNA | AY070728.1 | - | 12 - 13 | 931 | 59 | |
| BT000839.1 | - | 11 - 12 | 803 | 59 | ||
More Information
Set Membership: |
|

MAB1
Genomic Map