Assay Details
| Gene Symbol: MAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124463.4 | NP_199898.1 | 13 - 14 | 1218 | 72 | |
| GenBank mRNA | AY070728.1 | - | 13 - 14 | 1053 | 72 | |
| BT000839.1 | - | 12 - 13 | 925 | 72 | ||
More Information
Set Membership: |
|

MAB1
Genomic Map