Assay Details
| Gene Symbol: MAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124463.4 | NP_199898.1 | 5 - 6 | 501 | 119 | |
| GenBank mRNA | AY070728.1 | - | 5 - 6 | 336 | 119 | |
| BT000839.1 | - | 4 - 5 | 208 | 119 | ||
More Information
Set Membership: |
|

MAB1
Genomic Map