Assay Details
| Gene Symbol: MAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124463.4 | NP_199898.1 | 8 - 9 | 778 | 68 | |
| GenBank mRNA | AY070728.1 | - | 8 - 9 | 613 | 68 | |
| BT000839.1 | - | 7 - 8 | 485 | 68 | ||
More Information
Set Membership: |
|

MAB1
Genomic Map