Assay Details
| Gene Symbol: ERD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124486.3 | NP_568750.1 | 5 - 6 | 1650 | 75 | |
| GenBank mRNA | AY035112.1 | - | 5 - 6 | 1336 | 75 | |
| AY133868.1 | - | 5 - 6 | 1267 | 75 | ||
| D17582.1 | - | 5 - 6 | 1381 | 75 | ||
More Information
Set Membership: |
|

ERD1
Genomic Map