Assay Details
| Gene Symbol: AT5G51150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124494.2 | NP_199928.1 | 7 - 8 | 1451 | 61 | |
| GenBank mRNA | AY045838.1 | - | 7 - 8 | 1380 | 61 | |
| AY091374.1 | - | 7 - 8 | 1330 | 61 | ||
More Information
Set Membership: |
|

AT5G51150
Genomic Map