Assay Details
| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 12 - 13 | 1366 | 75 | |
| GenBank mRNA | AF216580.1 | - | 12 - 13 | 1081 | 75 | |
| AY099708.1 | - | 12 - 13 | 1037 | 75 | ||
| AY128901.1 | - | 12 - 13 | 972 | 75 | ||
More Information
Set Membership: |
|

PGM
Genomic Map