Assay Details
| Gene Symbol: PGM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124561.3 | NP_199995.1 | 6 - 7 | 911 | 100 | |
| GenBank mRNA | AF216580.1 | - | 6 - 7 | 626 | 100 | |
| AY099708.1 | - | 6 - 7 | 582 | 100 | ||
| AY128901.1 | - | 6 - 7 | 517 | 100 | ||
More Information
Set Membership: |
|

PGM
Genomic Map