Assay Details
| Gene Symbol: AGL72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085276.1 | NP_001078745.1 | 3 - 4 | 414 | 97 | |
| NM_124565.2 | NP_199999.1 | 3 - 4 | 414 | 97 | ||
| GenBank mRNA | AY141221.1 | - | 2 - 3 | 267 | 97 | |
| DQ447061.1 | - | 2 - 3 | 267 | 97 | ||
More Information
Set Membership: |
|

AGL72
Genomic Map