Assay Details
| Gene Symbol: OVA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124631.4 | NP_200065.1 | 4 - 5 | 674 | 64 | |
| GenBank mRNA | AK176421.1 | - | 4 - 5 | 631 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OVA6| Gene Symbol: OVA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124631.4 | NP_200065.1 | 4 - 5 | 674 | 64 | |
| GenBank mRNA | AK176421.1 | - | 4 - 5 | 631 | 64 | |
Set Membership: |
|