Assay Details
| Gene Symbol: PKP-BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124670.3 | NP_200104.1 | 11 - 12 | 1839 | 83 | |
| GenBank mRNA | AY084507.1 | - | 11 - 12 | 1763 | 83 | |
More Information
Set Membership: |
|

PKP-BETA1
Genomic Map