Assay Details
| Gene Symbol: PKP-BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124670.3 | NP_200104.1 | 4 - 5 | 1046 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PKP-BETA1| Gene Symbol: PKP-BETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124670.3 | NP_200104.1 | 4 - 5 | 1046 | 86 | |
Set Membership: |
|