Assay Details
| Gene Symbol: MLO11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036993.2 | NP_001032070.1 | 14 - 15 | 1727 | 116 | |
| NM_124755.3 | NP_200187.1 | 13 - 14 | 1817 | 116 | ||
| GenBank mRNA | AF369572.1 | - | 12 - 13 | 1176 | 116 | |
| AK317279.1 | - | 14 - 15 | 1517 | 116 | ||
| AY057502.1 | - | 13 - 14 | 1661 | 116 | ||
More Information
Set Membership: |
|

MLO11
Genomic Map