Assay Details
| Gene Symbol: MLO11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036993.2 | NP_001032070.1 | 15 - 16 | 1789 | 106 | |
| NM_124755.3 | NP_200187.1 | 14 - 15 | 1879 | 106 | ||
| GenBank mRNA | AF369572.1 | - | 13 - 14 | 1238 | 106 | |
| AK317279.1 | - | 15 - 16 | 1579 | 106 | ||
| AY057502.1 | - | 14 - 15 | 1723 | 106 | ||
| BT000434.1 | - | 14 - 15 | 1713 | 106 | ||
| BT002581.1 | - | 13 - 14 | 1238 | 106 | ||
More Information
Set Membership: |
|

MLO11
Genomic Map