Assay Details
| Gene Symbol: MLO11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036993.2 | NP_001032070.1 | 16 - 17 | 1851 | 101 | |
| NM_124755.3 | NP_200187.1 | 15 - 16 | 1941 | 101 | ||
| GenBank mRNA | AF369572.1 | - | 14 - 15 | 1300 | 101 | |
| AK317279.1 | - | 16 - 17 | 1641 | 101 | ||
| AY057502.1 | - | 15 - 16 | 1785 | 101 | ||
| BT000434.1 | - | 15 - 16 | 1775 | 101 | ||
| BT002581.1 | - | 14 - 15 | 1300 | 101 | ||
More Information
Set Membership: |
|

MLO11
Genomic Map