Assay Details
| Gene Symbol: MLO11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036993.2 | NP_001032070.1 | 4 - 5 | 760 | 135 | |
| NM_124755.3 | NP_200187.1 | 3 - 4 | 850 | 135 | ||
| GenBank mRNA | AF369572.1 | - | 2 - 3 | 209 | 135 | |
| AK317279.1 | - | 4 - 5 | 550 | 135 | ||
| AY057502.1 | - | 3 - 4 | 694 | 135 | ||
| BT000434.1 | - | 3 - 4 | 684 | 135 | ||
| BT002581.1 | - | 2 - 3 | 209 | 135 | ||
More Information
Set Membership: |
|

MLO11
Genomic Map