Assay Details
| Gene Symbol: AT2G37890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129345.4 | NP_181325.2 | 1 - 2 | 488 | 64 | |
| GenBank mRNA | AK175138.1 | - | 1 - 2 | 328 | 64 | |
| AK175263.1 | - | 1 - 2 | 336 | 64 | ||
| AY128317.1 | - | 1 - 2 | 370 | 64 | ||
| BT000042.1 | - | 1 - 2 | 209 | 64 | ||
More Information
Set Membership: |
|

AT2G37890
Genomic Map