Assay Details
| Gene Symbol: CYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202784.1 | NP_001189713.1 | 2 - 3 | 277 | 81 | |
| NM_001336797.1 | NP_001325272.1 | 2 - 3 | 347 | 81 | ||
| NM_129535.4 | NP_181507.1 | 2 - 3 | 417 | 81 | ||
| GenBank mRNA | AF076484.1 | - | 2 - 3 | 262 | 81 | |
| AF108660.1 | - | 2 - 3 | 260 | 81 | ||
| AF361812.1 | - | 2 - 3 | 268 | 81 | ||
| AF428297.1 | - | 2 - 3 | 268 | 81 | ||
| AY057541.1 | - | 2 - 3 | 268 | 81 | ||
| AY087698.1 | - | 2 - 3 | 264 | 81 | ||
| AY133643.1 | - | 1 - 2 | 110 | 81 | ||
| BT000697.1 | - | 2 - 3 | 268 | 81 | ||
| BT006365.1 | - | 1 - 2 | 110 | 81 | ||
More Information
Set Membership: |
|

CYT1
Genomic Map