Assay Details
| Gene Symbol: CYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202784.1 | NP_001189713.1 | 4 - 5 | 582 | 137 | |
| NM_001336797.1 | NP_001325272.1 | 4 - 5 | 652 | 137 | ||
| NM_129535.4 | NP_181507.1 | 4 - 5 | 722 | 137 | ||
| GenBank mRNA | AF076484.1 | - | 4 - 5 | 567 | 137 | |
| AF108660.1 | - | 4 - 5 | 565 | 137 | ||
| AF361812.1 | - | 4 - 5 | 573 | 137 | ||
| AF428297.1 | - | 4 - 5 | 573 | 137 | ||
| AY057541.1 | - | 4 - 5 | 573 | 137 | ||
| AY087698.1 | - | 4 - 5 | 569 | 137 | ||
| AY133643.1 | - | 3 - 4 | 415 | 137 | ||
| BT000697.1 | - | 4 - 5 | 573 | 137 | ||
| BT006365.1 | - | 3 - 4 | 415 | 137 | ||
More Information
Set Membership: |
|

CYT1
Genomic Map