Assay Details
| Gene Symbol: KT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084563.2 | NP_001078032.1 | 1 - 2 | 760 | 70 | |
| NM_001336844.1 | NP_001318392.1 | 2 - 3 | 249 | 70 | ||
| NM_001336845.1 | NP_001324081.1 | 2 - 3 | 689 | 70 | ||
| NM_001336846.1 | NP_001324082.1 | 2 - 3 | 666 | 70 | ||
| NM_129616.4 | NP_565936.1 | 2 - 3 | 425 | 70 | ||
| GenBank mRNA | BT004049.1 | - | 2 - 3 | 217 | 70 | |
| BT005048.1 | - | 1 - 2 | 47 | 70 | ||
More Information
Set Membership: |
|

KT2
Genomic Map