Assay Details
| Gene Symbol: KT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084563.2 | NP_001078032.1 | 4 - 5 | 1300 | 90 | |
| NM_001336844.1 | NP_001318392.1 | 5 - 6 | 789 | 90 | ||
| NM_001336845.1 | NP_001324081.1 | 5 - 6 | 1229 | 90 | ||
| NM_001336846.1 | NP_001324082.1 | 5 - 6 | 1206 | 90 | ||
| NM_129616.4 | NP_565936.1 | 5 - 6 | 965 | 90 | ||
| GenBank mRNA | AF012657.1 | - | 4 - 5 | 590 | 90 | |
| BT004049.1 | - | 5 - 6 | 757 | 90 | ||
| BT005048.1 | - | 4 - 5 | 587 | 90 | ||
More Information
Set Membership: |
|

KT2
Genomic Map