Assay Details
| Gene Symbol: TTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129819.3 | NP_565976.1 | 5 - 6 | 2191 | 93 | |
| GenBank mRNA | AF367321.1 | - | 5 - 6 | 1955 | 93 | |
| AY143941.1 | - | 5 - 6 | 1815 | 93 | ||
More Information
Set Membership: |
|

TTL3
Genomic Map