Assay Details
| Gene Symbol: ETFQO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337026.1 | NP_001324650.1 | 2 - 3 | 659 | 111 | |
| NM_129901.4 | NP_181868.1 | 2 - 3 | 696 | 111 | ||
More Information
Set Membership: |
|

ETFQO
Genomic Map