Assay Details
| Gene Symbol: AT4G11655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340725.1 | NP_001329008.1 | 1 - 2 | 393 | 91 | |
| NM_202806.2 | NP_974535.1 | 1 - 2 | 445 | 91 | ||
More Information
Set Membership: |
|

AT4G11655
Genomic Map