Assay Details
| Gene Symbol: AT4G11655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202806.2 | NP_974535.1 | 2 - 3 | 587 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G11655| Gene Symbol: AT4G11655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202806.2 | NP_974535.1 | 2 - 3 | 587 | 71 | |
Set Membership: |
|