Assay Details
| Gene Symbol: MEE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331621.1 | NP_001321282.1 | 4 - 5 | 2215 | 141 | |
| NM_100503.5 | NP_172112.1 | 4 - 5 | 2175 | 141 | ||
| NM_179269.4 | NP_849600.1 | 5 - 6 | 2308 | 141 | ||
| GenBank mRNA | AK226591.1 | - | 4 - 5 | 2163 | 141 | |
More Information
Set Membership: |
|

MEE5
Genomic Map