Assay Details
| Gene Symbol: MEE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331621.1 | NP_001321282.1 | 5 - 6 | 2318 | 133 | |
| NM_100503.5 | NP_172112.1 | 5 - 6 | 2278 | 133 | ||
| NM_179269.4 | NP_849600.1 | 6 - 7 | 2411 | 133 | ||
| GenBank mRNA | AK226591.1 | - | 5 - 6 | 2266 | 133 | |
More Information
Set Membership: |
|

MEE5
Genomic Map