Assay Details
| Gene Symbol: AGL51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202771.1 | NP_974500.1 | 1 - 2 | 221 | 158 | |
| GenBank mRNA | AY141215.1 | - | 1 - 2 | 221 | 158 | |
More Information
Set Membership: |
|

AGL51
Genomic Map