Assay Details
| Gene Symbol: LPA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105964.5 | NP_565054.1 | 7 - 8 | 746 | 155 | |
| GenBank mRNA | BT002009.1 | - | 7 - 8 | 698 | 155 | |
| BT008843.1 | - | 7 - 8 | 689 | 155 | ||
More Information
Set Membership: |
|

LPA3
Genomic Map