Assay Details
| Gene Symbol: FUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334648.1 | NP_001320577.1 | 1 - 2 | 421 | 151 | |
| NM_106102.4 | NP_177582.3 | 1 - 2 | 335 | 151 | ||
| GenBank mRNA | AK175419.1 | - | 1 - 2 | 338 | 151 | |
| AK176710.1 | - | 1 - 2 | 338 | 151 | ||
| KJ138935.1 | - | 1 - 2 | 255 | 151 | ||
More Information
Set Membership: |
|

FUT3
Genomic Map