Assay Details
| Gene Symbol: FBA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341806.1 | NP_001329762.1 | 1 - 2 | 750 | 146 | |
| NM_001341807.1 | NP_001329763.1 | 2 - 3 | 612 | 146 | ||
| NM_001341808.1 | NP_001329761.1 | 1 - 2 | 321 | 146 | ||
| NM_118785.4 | NP_194382.1 | 2 - 3 | 612 | 146 | ||
| GenBank mRNA | BT001927.1 | - | 2 - 3 | 284 | 146 | |
More Information
Set Membership: |
|

FBA7
Genomic Map