Assay Details
| Gene Symbol: AT5G03820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342734.1 | NP_001318474.1 | 5 - 6 | 1049 | 134 | |
| NM_120463.3 | NP_196002.2 | 4 - 5 | 1133 | 134 | ||
More Information
Set Membership: |
|

AT5G03820
Genomic Map