Assay Details
| Gene Symbol: RPL5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203510.1 | NP_001190439.1 | 6 - 7 | 600 | 141 | |
| NM_123336.6 | NP_198790.1 | 5 - 6 | 575 | 141 | ||
| GenBank mRNA | AY079020.1 | - | 5 - 6 | 531 | 141 | |
| AY093123.1 | - | 5 - 6 | 519 | 141 | ||
| BT008706.1 | - | 4 - 5 | 489 | 141 | ||
More Information
Set Membership: |
|

RPL5B
Genomic Map