Assay Details
| Gene Symbol: SYP111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100729.4 | NP_172332.1 | 2 | 1127 | 75 | |
| GenBank mRNA | U39451.1 | - | 2 | 1038 | 75 | |
| Gene Symbol: NPQ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331777.1 | NP_001321301.1 | 5 | 2380 | 75 | |
| NM_100728.4 | NP_172331.1 | 6 | 2085 | 75 | ||
More Information
Set Membership: |
|

SYP111
Genomic Map