Assay Details
| Gene Symbol: RANGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203237.1 | NP_001190166.1 | 2 | 1448 | 143 | |
| NM_116178.3 | NP_191872.1 | 2 | 1581 | 143 | ||
| GenBank mRNA | AF214559.1 | - | 1 | 1205 | 143 | |
| AY034918.1 | - | 2 | 1441 | 143 | ||
| AY150448.1 | - | 1 | 1205 | 143 | ||
More Information
Set Membership: |
|

RANGAP1
Genomic Map