Assay Details
| Gene Symbol: AT5G07330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120815.3 | NP_196350.1 | 2 | 705 | 143 | |
| GenBank mRNA | AK118558.2 | - | 2 | 672 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G07330| Gene Symbol: AT5G07330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120815.3 | NP_196350.1 | 2 | 705 | 143 | |
| GenBank mRNA | AK118558.2 | - | 2 | 672 | 143 | |
Set Membership: |
|