Assay Details
| Gene Symbol: ATSLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127354.5 | NP_179389.1 | 2 | 2044 | 79 | |
| GenBank mRNA | AY050414.1 | - | 2 | 2035 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATSLY1| Gene Symbol: ATSLY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127354.5 | NP_179389.1 | 2 | 2044 | 79 | |
| GenBank mRNA | AY050414.1 | - | 2 | 2035 | 79 | |
Set Membership: |
|